missing translation for 'onlineSavingsMsg'
Learn More

Natamycin Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111015473
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015473 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015473 Supplier Merck Emd Millipore Supplier No. PHR17031G
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.Natamycin is a major drug that is utilized in the treatment of fungal keratitis. It is, by far, the only available approved antifungal drug for the said purpose by the U.S. Food and Drug Administration.

Specifications

CAS 7681-93-8
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C33H47 NO13
MDL Number MFCD00135085
Synonym Pimaricin; Natamycin
Type Certified Reference Material
Quantity 1 g
Formula Weight 665.73
Grade Certified Reference Material