missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Nasco Whirl-Pak™ Standard Sample Bags

Features puncture proof tabs for protection from damage due to wire-end protrusion. Nasco™ Whirl-Pak™ Easy-To-Close Bags are ideal for sampling and product analysis.

Manufacturer:  Nasco B01323WA

 View more versions of this product

Catalog No. 01-812-3




Description

Description

Features puncture proof tabs for protection from damage due to wire-end protrusion. Nasco™ Whirl-Pak™ Easy-To-Close Bags are ideal for sampling and product analysis.

  • Transparent polyethylene bags
  • Gas sterilized, with documentation available
  • Pull tabs help prevent contamination from finger contact when opening
  • Write-on bags have area that accepts ballpoint pen or pencil
  • Bags can be frozen to any temperature; including liquid nitrogen--careful handling required after freezing
  • Thickness, 2.25 to 4 mil

Bags should not be used at temperatures above 180°F (82°C).

Specifications

Specifications

Plain Style
3 mil
2.04 L
Transparent
PE (Polyethylene)
19 cm
38 x 19 cm
69 oz.
Closing Tab
38 cm
0.076 mm
SDS
Documents