missing translation for 'onlineSavingsMsg'
Learn More

Eisco™ Reagent Bottles

Catalog No. S24489
Change view
Click to view available options
Mouth:
Wide
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Mouth
S24489 Wide
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S24489 Supplier Eisco™ Supplier No. CH0172DW

Rigid bottles for universities and industrial labs

  • Made from polypropylene
  • Available as wide or narrow neck
  • Autoclavable
  • Leak-proof

Specifications

Capacity (Metric) 500 mL
Capacity (English) 17 oz.
Color Clear
Description Leak-proof
Material Polypropylene
Length (Metric) 6.98 cm
Quantity 1 Ea.
Sterility Non-sterile
Width (Metric) 6.98 cm
Closure Included Yes
Graduated No
Mouth Wide
Height (Metric) 17.14 cm
Product Type Reagent Bottle
Height (English) 6.75 in.
Autoclavable Autoclavable
Show More Show Less