missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eisco Labs Narrow Mouth Polypropylene Reagent Bottles with Polypropylene Caps
Description
- Narrow Mouth Bottles: Translucent polypropylene reagent bottles with polypropylene caps, designed for safe and efficient liquid storage in labs and industrial settings.
- Temperature And Chemical Resistance: Operates in a range of 0-135 °C and resists various chemicals, making it suitable for diverse lab and storage needs.
- Leak-Proof Design: Rigorously tested to prevent leaks during transport or storage. Features semi-buttress threads and secure polypropylene caps for tight closure.
- Autoclavable And Non-Sterile: Ideal for reusable lab workflows. Made from FDA-compliant and BSE/TSE-free materials.
- Lab And Industrial Applications: Suitable for research, sample storage, and reagent mixing. Translucent body provides visibility, and design ensures easy transport and handling.
Specifications
Specifications
| Capacity (Metric) | 125 mL |
| Capacity (English) | 4.22 oz. |
| Color | Translucent |
| Material | Polypropylene |
| No. per Pack | 12 Pk. |
| Sterility | Non-sterile |
| Closure Included | Yes |
| Graduated | No |
| Mouth | Narrow |
| Height (Metric) | 100 mm |
| Show More |