missing translation for 'onlineSavingsMsg'
Learn More

Eisco Labs Narrow Mouth Polypropylene Reagent Bottles with Polypropylene Caps

Translucent polypropylene narrow mouth bottles with matching polypropylene caps offer chemical resistance, leakproof performance, and easy handling - perfect for safe liquid storage and transport in lab and industrial environments.

Supplier:  Eisco Labs CH5338PK12

 View more versions of this product

Catalog No. S000071

Only null left

Description

Description

  • Narrow Mouth Bottles: Translucent polypropylene reagent bottles with polypropylene caps, designed for safe and efficient liquid storage in labs and industrial settings.
  • Temperature And Chemical Resistance: Operates in a range of 0-135 °C and resists various chemicals, making it suitable for diverse lab and storage needs.
  • Leak-Proof Design: Rigorously tested to prevent leaks during transport or storage. Features semi-buttress threads and secure polypropylene caps for tight closure.
  • Autoclavable And Non-Sterile: Ideal for reusable lab workflows. Made from FDA-compliant and BSE/TSE-free materials.
  • Lab And Industrial Applications: Suitable for research, sample storage, and reagent mixing. Translucent body provides visibility, and design ensures easy transport and handling.
Specifications

Chemical Identifiers

Translucent
12 Pk.
Yes
Narrow
Autoclavable
Polypropylene
Non-sterile
No
Reagent Bottle

Specifications

15 mL
Translucent
12 Pk.
Yes
Narrow
Reagent Bottle
Autoclavable
0.50 oz.
Polypropylene
Non-sterile
No
58 mm
25 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions