missing translation for 'onlineSavingsMsg'
Learn More

Eisco Labs Narrow Mouth Polypropylene Reagent Bottles with Polypropylene Caps

Catalog No. S000070
Change view
Click to view available options
Capacity (Metric):
8 mL
15 mL
60 mL
125 mL
500 mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
S000070 8 mL
S000071 15 mL
S000072 60 mL
S000073 125 mL
S000074 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. S000070 Supplier Eisco Labs Supplier No. CH5337PK12

Translucent polypropylene narrow mouth bottles with matching polypropylene caps offer chemical resistance, leakproof performance, and easy handling - perfect for safe liquid storage and transport in lab and industrial environments.

  • Narrow Mouth Bottles: Translucent polypropylene reagent bottles with polypropylene caps, designed for safe and efficient liquid storage in labs and industrial settings.
  • Temperature And Chemical Resistance: Operates in a range of 0-135 °C and resists various chemicals, making it suitable for diverse lab and storage needs.
  • Leak-Proof Design: Rigorously tested to prevent leaks during transport or storage. Features semi-buttress threads and secure polypropylene caps for tight closure.
  • Autoclavable And Non-Sterile: Ideal for reusable lab workflows. Made from FDA-compliant and BSE/TSE-free materials.
  • Lab And Industrial Applications: Suitable for research, sample storage, and reagent mixing. Translucent body provides visibility, and design ensures easy transport and handling.

Specifications

Capacity (Metric) 8 mL
Capacity (English) 0.27 oz.
Color Translucent
Material Polypropylene
No. per Pack 12 Pk.
Sterility Non-sterile
Closure Included Yes
Graduated No
Mouth Narrow
Height (Metric) 45 mm
Product Type Reagent Bottle
Diameter (Metric) 25 mm
Autoclavable Autoclavable
Show More Show Less