missing translation for 'onlineSavingsMsg'
Learn More

Eisco Labs Narrow Mouth Amber HDPE Reagent Bottles with Polypropylene Caps

Narrow-mouth amber HDPE reagent bottles with polypropylene caps offers UV protection, leakproof performance, and chemical resistance that are ideal for safely storing light-sensitive liquids in lab or industrial settings.

Supplier:  Eisco Labs CH5380PK12

 View more versions of this product

Catalog No. S000091


Only null left

Description

Description

  • Amber Reagent Bottles: Includes narrow-mouth HDPE reagent bottles with polypropylene caps, ideal for storing and transporting light-sensitive substances.
  • Protect Light-Sensitive Samples: Amber color shields UV-sensitive reagents and samples, perfect for laboratory or industrial use.
  • Leak-Proof and Secure: Rigorously tested for leak resistance under high-pressure conditions to ensure safe and reliable storage.
  • Excellent Chemical Resistance: HDPE construction withstands a variety of chemicals, making it a versatile choice for lab applications.
  • Durable and Easy To Handle: Lightweight and translucent designed for convenience in handling and transport.
Specifications

Specifications

1 L
Amber
HDPE
Non-sterile
No
219 mm
97 mm
33.8 oz.
28.8 mm
12 Pk.
Yes
Narrow
Reagent Bottle
Not Autoclavable
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions