missing translation for 'onlineSavingsMsg'
Learn More

Naringin, ≥95% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111012310
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012310 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012310 Supplier Merck Emd Millipore Supplier No. 91842500MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Naringin is a naturally occurring flavone glycoside, which may possess many pharmacological properties like anti-inflammatory, anti-oxidant and anti-apoptotic activity. It is widely found in fruits like grapes and oranges.

Specifications

CAS 10236-47-2
Physical Form Neat
Linear Formula C27H32O14
MDL Number MFCD00148888
Synonym 4′,5,7-Trihydroxyflavanone 7-rhamnoglucoside; Naringenin 7-neohesperidoside; Naringenine-7-rhamnosidoglucoside; Naringoside
Type Analytical Standard
Quantity 500 mg
Formula Weight 580.53
Percent Purity ≥95% (HPLC)
Grade Analytical Standard