missing translation for 'onlineSavingsMsg'
Learn More

(±)-Naringenin, ≥95% (TLC), MilliporeSigma™ Supelco™

Catalog No. 111015138
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015138 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015138 Supplier Merck Emd Millipore Supplier No. 5218650MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

(+/-)-Naringenin is a naturally occurring plant bioflavonoid present in orange and grape type of citrus fruit juices, which can exhibit pharmacological properties like anti-carcinogenic, estrogenic and anti-oxidative.

Featured Industries: Food and Beverages

Specifications

CAS 67604-48-2
Physical Form Neat
Linear Formula C15H12O5
MDL Number MFCD00006844
Melting Point 247°C to 250°C
Synonym (+/-)-2,3-Dihydro-5,7-dihydroxy-2-(4-hydroxyphenyl)-4 H-1-benzopyran-4-one; 4′,5,7-Trihydroxyflavanone
Type Analytical Standard
Quantity 50 mg
Formula Weight 272.25
Percent Purity ≥95% (TLC)
Grade Analytical Standard
Show More Show Less