missing translation for 'onlineSavingsMsg'
Learn More

Naproxen sodium Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111013771
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013771 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013771 Supplier Merck Emd Millipore Supplier No. PHR11651G
Naproxen sodium (NS) is a sodium salt of propionic acid derivative and demonstrates analgesic, anti-pyretic, and anti-inflammatory properties. It exhibits efficiency in being used as an indomethacin in the treatment of acute musculoskeletal disorders. It belongs to the group of nonsteroidal anti-inflammatory drugs (NSAIDS) and acts by inhibiting the activity of both the cyclooxygenase enzymes, COX-1 & COX-2 and further blocking the synthesis of certain prostaglandins. It is administered in the treatment of ankylosing spondylitis, osteoarthritis, rheumatoid disorders, acute gout, mild to moderate pain, tendonitis, bursitis, dysmenorrhea, fever, and migraine headache. Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Specifications

CAS 26159-34-2
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Physical Form Neat
Linear Formula C14H13 NaO3
MDL Number MFCD00058507
Synonym (S)-6-Methoxy-alpha-methyl-2-naphthaleneacetic acid sodium salt
Type Certified Reference Material
Quantity 1 g
Formula Weight 252.24
Grade Certified Reference Material