missing translation for 'onlineSavingsMsg'
Learn More

Naproxen Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111019310
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019310 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019310 Supplier Merck Emd Millipore Supplier No. PHR1040500MG
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.Naproxen belongs to the class of arylacetic acids and is used as a non-steroidal anti-inflammatory drug (NSAID). It is useful for the management of pain, inflammation and stiffness. It is also used in the treatment of rheumatoid arthritis and other painful musculoskeletal disorders.

Specifications

CAS 22204-53-1
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Physical Form Neat
Linear Formula CH3 OC10H6 CH(CH3)CO2 H
MDL Number MFCD00010500
Melting Point 152°C to 154°C
Synonym (S)-(+)-2-(6-Methoxy-2-naphthyl)propionic acid
Type Certified Reference Material
Quantity 500 mg
Formula Weight 230.26
Grade Certified Reference Material
Show More Show Less