missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Naphthol Green B, ACS Reagent CI 10020, Reagents
Naphthol Green B, ACS Reagent CI 10020, ACS, for general laboratory use. Manufactured in ISO 9001 facility.
$534.23 - $1,869.40
Chemical Identifiers
| CAS | 19381-50-1 |
|---|---|
| Molecular Formula | C30H18FeN3Na3O15S3 |
| Molecular Weight (g/mol) | 881.47 |
| InChI Key | VLBLPLXOYXEXJK-UHFFFAOYSA-K |
| Synonym | Acid Green 1, Color Index 10020 |
| IUPAC Name | trisodium tris(6-hydroxy-5-nitrosonaphthalene-2-sulfonate) iron |
| SMILES | [Na+].[Na+].[Na+].[Fe].OC1=C(N=O)C2=CC=C(C=C2C=C1)S([O-])(=O)=O.OC1=C(N=O)C2=CC=C(C=C2C=C1)S([O-])(=O)=O.OC1=C(N=O)C2=CC=C(C=C2C=C1)S([O-])(=O)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
01-337-164
|
Labchem Inc
C2252400100A6 |
100 mL |
N/A
|
|
|||||
|
01-337-163
|
Labchem Inc
C2252400500A5 |
500 g |
N/A
|
|
|||||
Chemical Identifiers
| 19381-50-1 | |
| 881.47 | |
| Acid Green 1, Color Index 10020 | |
| [Na+].[Na+].[Na+].[Fe].OC1=C(N=O)C2=CC=C(C=C2C=C1)S([O-])(=O)=O.OC1=C(N=O)C2=CC=C(C=C2C=C1)S([O-])(=O)=O.OC1=C(N=O)C2=CC=C(C=C2C=C1)S([O-])(=O)=O |
| C30H18FeN3Na3O15S3 | |
| VLBLPLXOYXEXJK-UHFFFAOYSA-K | |
| trisodium tris(6-hydroxy-5-nitrosonaphthalene-2-sulfonate) iron |
Specifications
| 2735°C | |
| C30H18FeN3Na3O15S3 | |
| VLBLPLXOYXEXJK-UHFFFAOYSA-K | |
| trisodium tris(6-hydroxy-5-nitrosonaphthalene-2-sulfonate) iron | |
| ACS | |
| 1 | |
| 9.5 | |
| Solid |
| 19381-50-1 | |
| Acid Green 1, Color Index 10020 | |
| [Na+].[Na+].[Na+].[Fe].OC1=C(N=O)C2=CC=C(C=C2C=C1)S([O-])(=O)=O.OC1=C(N=O)C2=CC=C(C=C2C=C1)S([O-])(=O)=O.OC1=C(N=O)C2=CC=C(C=C2C=C1)S([O-])(=O)=O | |
| 881.47 | |
| Poly Bottle | |
| Black/Green | |
| 100 mL | |
| Naphthol Green B, ACS Reagent CI 10020 |