missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Naphthol Green B, ACS Reagent CI 10020, Reagents
Naphthol Green B, ACS Reagent CI 10020, ACS, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Labchem Inc C2252400500A5
Specifications
| Naphthol Green B, ACS Reagent CI 10020 | |
| 19381-50-1 | |
| Acid Green 1, Color Index 10020 | |
| [Na+].[Na+].[Na+].[Fe].OC1=C(N=O)C2=CC=C(C=C2C=C1)S([O-])(=O)=O.OC1=C(N=O)C2=CC=C(C=C2C=C1)S([O-])(=O)=O.OC1=C(N=O)C2=CC=C(C=C2C=C1)S([O-])(=O)=O | |
| 881.47 | |
| Poly Bottle | |
| Black/Green | |
| 500 g |
| 2735°C | |
| C30H18FeN3Na3O15S3 | |
| VLBLPLXOYXEXJK-UHFFFAOYSA-K | |
| trisodium tris(6-hydroxy-5-nitrosonaphthalene-2-sulfonate) iron | |
| ACS | |
| 1 | |
| 9.5 | |
| Solid |
Chemical Identifiers
| 19381-50-1 | |
| 881.47 | |
| Acid Green 1, Color Index 10020 | |
| [Na+].[Na+].[Na+].[Fe].OC1=C(N=O)C2=CC=C(C=C2C=C1)S([O-])(=O)=O.OC1=C(N=O)C2=CC=C(C=C2C=C1)S([O-])(=O)=O.OC1=C(N=O)C2=CC=C(C=C2C=C1)S([O-])(=O)=O |
| C30H18FeN3Na3O15S3 | |
| VLBLPLXOYXEXJK-UHFFFAOYSA-K | |
| trisodium tris(6-hydroxy-5-nitrosonaphthalene-2-sulfonate) iron |