missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Naphthol Green B, pure
CAS: 19381-50-1 | C30H18FeN3Na3O15S3 | 881.474 g/mol
$53.21 - $469.09
Chemical Identifiers
| CAS | 19381-50-1 |
|---|---|
| Molecular Formula | C30H18FeN3Na3O15S3 |
| Molecular Weight (g/mol) | 881.474 |
| MDL Number | MFCD00003886 |
| InChI Key | VLBLPLXOYXEXJK-UHFFFAOYSA-K |
| Synonym | Acid Green 1, C.I. 10020 |
| PubChem CID | 14598749 |
| IUPAC Name | trisodium;6-hydroxy-5-nitrosonaphthalene-2-sulfonate;iron |
| SMILES | C1=CC2=C(C=CC(=C2N=O)O)C=C1S(=O)(=O)[O-].C1=CC2=C(C=CC(=C2N=O)O)C=C1S(=O)(=O)[O-].C1=CC2=C(C=CC(=C2N=O)O)C=C1S(=O)(=O)[O-].[Na+].[Na+].[Na+].[Fe] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC152930250
|
Thermo Scientific Chemicals
152930250 |
25 g | Glass bottle |
Each for $53.21
|
|
||||
|
AC152935000
|
Thermo Scientific Chemicals
152935000 |
500 g | Glass bottle |
Each for $469.09
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
IndicatorChemical Identifiers
| 19381-50-1 | |
| 881.474 | |
| VLBLPLXOYXEXJK-UHFFFAOYSA-K | |
| 14598749 | |
| C1=CC2=C(C=CC(=C2N=O)O)C=C1S(=O)(=O)[O-].C1=CC2=C(C=CC(=C2N=O)O)C=C1S(=O)(=O)[O-].C1=CC2=C(C=CC(=C2N=O)O)C=C1S(=O)(=O)[O-].[Na+].[Na+].[Na+].[Fe] |
| C30H18FeN3Na3O15S3 | |
| MFCD00003886 | |
| Acid Green 1, C.I. 10020 | |
| trisodium;6-hydroxy-5-nitrosonaphthalene-2-sulfonate;iron |
Specifications
| 19381-50-1 | |
| MFCD00003886 | |
| Acid Green 1, C.I. 10020 | |
| VLBLPLXOYXEXJK-UHFFFAOYSA-K | |
| C1=CC2=C(C=CC(=C2N=O)O)C=C1S(=O)(=O)[O-].C1=CC2=C(C=CC(=C2N=O)O)C=C1S(=O)(=O)[O-].C1=CC2=C(C=CC(=C2N=O)O)C=C1S(=O)(=O)[O-].[Na+].[Na+].[Na+].[Fe] | |
| 881.474 | |
| 878.47 | |
| Glass bottle | |
| Green | |
| Crystalline Powder |
| C30H18FeN3Na3O15S3 | |
| 11, 332 | |
| Solubility in water: 30 mg/mL water. Other solubilities: 0.9 mg/mL ethanol, 9 mg/mL ethylene glycol monomethyl ether | |
| Authentic | |
| trisodium;6-hydroxy-5-nitrosonaphthalene-2-sulfonate;iron | |
| 14598749 | |
| Pure | |
| Complies | |
| 25 g | |
| Naphthol Green B |
RUO – Research Use Only