missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Naphthol Green B, pure
CAS: 19381-50-1 | C30H18FeN3Na3O15S3 | 881.474 g/mol
Supplier: Thermo Scientific Chemicals 152935000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
IndicatorSpecifications
| Naphthol Green B | |
| C30H18FeN3Na3O15S3 | |
| 11, 332 | |
| Solubility in water: 30 mg/mL water. Other solubilities: 0.9 mg/mL ethanol, 9 mg/mL ethylene glycol monomethyl ether | |
| Authentic | |
| trisodium;6-hydroxy-5-nitrosonaphthalene-2-sulfonate;iron | |
| 14598749 | |
| Pure | |
| Complies | |
| 500 g |
| 19381-50-1 | |
| MFCD00003886 | |
| Acid Green 1, C.I. 10020 | |
| VLBLPLXOYXEXJK-UHFFFAOYSA-K | |
| C1=CC2=C(C=CC(=C2N=O)O)C=C1S(=O)(=O)[O-].C1=CC2=C(C=CC(=C2N=O)O)C=C1S(=O)(=O)[O-].C1=CC2=C(C=CC(=C2N=O)O)C=C1S(=O)(=O)[O-].[Na+].[Na+].[Na+].[Fe] | |
| 881.474 | |
| 878.47 | |
| Glass bottle | |
| Green | |
| Crystalline Powder |
Chemical Identifiers
| 19381-50-1 | |
| 881.474 | |
| VLBLPLXOYXEXJK-UHFFFAOYSA-K | |
| 14598749 | |
| C1=CC2=C(C=CC(=C2N=O)O)C=C1S(=O)(=O)[O-].C1=CC2=C(C=CC(=C2N=O)O)C=C1S(=O)(=O)[O-].C1=CC2=C(C=CC(=C2N=O)O)C=C1S(=O)(=O)[O-].[Na+].[Na+].[Na+].[Fe] |
| C30H18FeN3Na3O15S3 | |
| MFCD00003886 | |
| Acid Green 1, C.I. 10020 | |
| trisodium;6-hydroxy-5-nitrosonaphthalene-2-sulfonate;iron |
RUO – Research Use Only