missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Naphthol AS-BI beta-D-Glucuronide 98.0+%, TCI America™
$100.11 - $325.36
Chemical Identifiers
| CAS | 37-87-6 |
|---|---|
| Molecular Formula | C24H22BrNO9 |
| Molecular Weight (g/mol) | 548.342 |
| MDL Number | MFCD00067162 |
| InChI Key | ACOOAEDFQKSZTC-NABGWTBKSA-N |
| Synonym | 6-Bromo-3[[(2-methoxyphenyl)amino]carbonyl]-2-naphthyl beta-D-Glucopyranosiduronic Acid |
| PubChem CID | 16219726 |
| IUPAC Name | (2S,3S,4S,5R,6S)-6-[6-bromo-3-[(2-methoxyphenyl)carbamoyl]naphthalen-2-yl]oxy-3,4,5-trihydroxyoxane-2-carboxylic acid |
| SMILES | COC1=CC=CC=C1NC(=O)C2=C(C=C3C=CC(=CC3=C2)Br)OC4C(C(C(C(O4)C(=O)O)O)O)O |
Chemical Identifiers
| 37-87-6 | |
| 548.342 | |
| ACOOAEDFQKSZTC-NABGWTBKSA-N | |
| 16219726 | |
| COC1=CC=CC=C1NC(=O)C2=C(C=C3C=CC(=CC3=C2)Br)OC4C(C(C(C(O4)C(=O)O)O)O)O |
| C24H22BrNO9 | |
| MFCD00067162 | |
| 6-Bromo-3[[(2-methoxyphenyl)amino]carbonyl]-2-naphthyl beta-D-Glucopyranosiduronic Acid | |
| (2S,3S,4S,5R,6S)-6-[6-bromo-3-[(2-methoxyphenyl)carbamoyl]naphthalen-2-yl]oxy-3,4,5-trihydroxyoxane-2-carboxylic acid |
Specifications
| 37-87-6 | |
| C24H22BrNO9 | |
| 25 mg | |
| ACOOAEDFQKSZTC-NABGWTBKSA-N | |
| (2S,3S,4S,5R,6S)-6-[6-bromo-3-[(2-methoxyphenyl)carbamoyl]naphthalen-2-yl]oxy-3,4,5-trihydroxyoxane-2-carboxylic acid | |
| 16219726 | |
| ≥98.0% (HPLC,N) | |
| Naphthol AS-BI beta-D-Glucuronide [for Biochemical Research] |
| White | |
| MFCD00067162 | |
| 6-Bromo-3[[(2-methoxyphenyl)amino]carbonyl]-2-naphthyl beta-D-Glucopyranosiduronic Acid | |
| COC1=CC=CC=C1NC(=O)C2=C(C=C3C=CC(=CC3=C2)Br)OC4C(C(C(C(O4)C(=O)O)O)O)O | |
| 548.342 | |
| 548.34 | |
| Crystalline Powder |
Safety and Handling
TSCA : No
Recommended Storage : Freezer