accessibility menu, dialog, popup

UserName

CAS RN 37-87-6

CH3OHNOBrOOOOHOHHOHO

CAS RN 37-87-6

IUPAC Name: 6-({6-bromo-3-[(2-methoxyphenyl)carbamoyl]naphthalen-2-yl}oxy)-3,4,5-trihydroxyoxane-2-carboxylic acid
Synonyms: 6-({6-bromo-3-[(2-methoxyphenyl)carbamoyl]naphthalen-2-yl}oxy)-3,4,5-trihydroxyoxane-2-carboxylic acid
Molecular Weight (g/mol): 548.34
Molecular Formula: C24H22BrNO9
InChi Key: ACOOAEDFQKSZTC-UHFFFAOYNA-N
SMILES: COC1=CC=CC=C1NC(=O)C1=C(OC2OC(C(O)C(O)C2O)C(O)=O)C=C2C=CC(Br)=CC2=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results