missing translation for 'onlineSavingsMsg'
Learn More

Naphtho[2,3-a]pyrene 98.0+%, TCI America™

Catalog No. N0599100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N0599100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N0599100MG Supplier TCI America Supplier No. N0599100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 196-42-9
Molecular Formula C24H14
Molecular Weight (g/mol) 302.376
MDL Number MFCD00191469
InChI Key AXSJLZJXXUBRBS-UHFFFAOYSA-N
PubChem CID 9137
SMILES C1=CC=C2C=C3C4=C5C(=CC3=CC2=C1)C=CC6=C5C(=CC=C6)C=C4

Specifications

Quantity 100 mg
Formula Weight 302.38
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Naphtho[2,3-a]pyrene