missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Naphtho[1,2-c:5,6-c']bis([1,2,5]thiadiazole) 98.0+%, TCI America™
Supplier: TCI America N1105200MG
Specifications
| Naphtho[1,2-c:5,6-c′]bis([1,2,5]thiadiazole) | |
| 248°C | |
| MFCD28579827 | |
| VHHDKHOBNKXVNU-UHFFFAOYSA-N | |
| [2,1,3]benzothiadiazolo[7,6-g][2,1,3]benzothiadiazole | |
| 14896996 | |
| ≥98.0% (HPLC) |
| 133546-47-1 | |
| C10H4N4S2 | |
| 200 mg | |
| C1=CC2=NSN=C2C3=C1C4=NSN=C4C=C3 | |
| 244.29 | |
| 244.29 | |
| Crystalline Powder |
Chemical Identifiers
| 133546-47-1 | |
| 244.29 | |
| VHHDKHOBNKXVNU-UHFFFAOYSA-N | |
| [2,1,3]benzothiadiazolo[7,6-g][2,1,3]benzothiadiazole |
| C10H4N4S2 | |
| MFCD28579827 | |
| 14896996 | |
| C1=CC2=NSN=C2C3=C1C4=NSN=C4C=C3 |
Safety and Handling
TSCA : No