accessibility menu, dialog, popup

UserName

CAS RN 133546-47-1

IUPAC Name: 4,12-dithia-3,5,11,13-tetraazatetracyclo[7.7.0.0²,⁶.0¹⁰,¹⁴]hexadeca-1(9),2,5,7,10,13,15-heptaene
Synonyms: 4,12-dithia-3,5,11,13-tetraazatetracyclo[7.7.0.0²,⁶.0¹⁰,¹⁴]hexadeca-1(9),2,5,7,10,13,15-heptaene
Molecular Weight (g/mol): 244.29
Molecular Formula: C10H4N4S2
InChi Key: VHHDKHOBNKXVNU-UHFFFAOYSA-N
SMILES: S1N=C2C=CC3=C(C=CC4=NSN=C34)C2=N1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results