missing translation for 'onlineSavingsMsg'
Learn More

Naphthenic Acid, TCI America™

Catalog No. p-7133758
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N034325G 25 g
N0343500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N034325G Supplier TCI America Supplier No. N034325G

Chemical Identifiers

CAS 1338-24-5
MDL Number MFCD00147696

Specifications

Quantity 25 g
UN Number 3082
Physical Form Liquid
Chemical Name or Material Naphthenic Acid
EINECSNumber (9)-1757
RTECSNumber QK8750000
TSCA Yes