CAS RN 1338-24-5
IUPAC Name:
3-(3-ethylcyclopentyl)propanoic acid
Synonyms:
3-(3-ethylcyclopentyl)propanoic acid
Molecular Weight (g/mol):
170.25
Molecular Formula:
C10H18O2
InChi Key:
WVRFSLWCFASCIS-UHFFFAOYNA-N
SMILES:
CCC1CCC(CCC(O)=O)C1
1
–
3
of
3
results
1
2
Naphthenic Acid, TCI America™
CAS: 1338-24-5 MDL Number: MFCD00147696
3
Sigma Aldrich Naphthenic acid
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.