missing translation for 'onlineSavingsMsg'
Learn More

Naphthenic Acid, TCI America™

Catalog No. N0343500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N0343500G 500 g
N034325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N0343500G Supplier TCI America Supplier No. N0343500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1338-24-5
MDL Number MFCD00147696

Specifications

Quantity 500 g
UN Number 3082
Physical Form Liquid
Chemical Name or Material Naphthenic Acid
EINECSNumber (9)-1757
RTECSNumber QK8750000
TSCA Yes