missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Naphthenic Acid, TCI America™
Specifications
Specifications
| Quantity | 25 g |
| UN Number | 3082 |
| Physical Form | Liquid |
| Chemical Name or Material | Naphthenic Acid |
Safety and Handling
| EINECSNumber | (9)-1757 |
| RTECSNumber | QK8750000 |
| TSCA | Yes |