missing translation for 'onlineSavingsMsg'
Learn More

Naphthalene-2-sulfonic acid, 98%

Catalog No. AAH5668522
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5668522 100 g
AAH5668536 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5668522 Supplier Thermo Scientific Chemicals Supplier No. H5668522

CAS: 120-18-3 | C10H8O3S | 208.23 g/mol

Naphthalene-2-sulfonic acid is used in making direct dyes, reactive des and employed as dye and agrochemical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Naphthalene-2-sulfonic acid is used in making direct dyes, reactive des and employed as dye and agrochemical intermediates.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with oxidizing agents. Store at room temperature.

Chemical Identifiers

CAS 120-18-3
Molecular Formula C10H8O3S
Molecular Weight (g/mol) 208.23
MDL Number MFCD00004089
InChI Key KVBGVZZKJNLNJU-UHFFFAOYSA-N
Synonym 2-naphthalenesulfonic acid, beta-naphthylsulfonic acid, beta-naphthalenesulfonic acid, naphthalene-2-sulphonic acid, kyselina 2-naftalensulfonova, 2-naphthalene sulfonic acid, unii-o9s4k2s25e, 2-naphthylsulfonic acid, kyselina 2-naftalensulfonova czech, 2-naphthalenesulfonic acid, monohydrate
PubChem CID 8420
ChEBI CHEBI:44229
IUPAC Name naphthalene-2-sulfonic acid
SMILES OS(=O)(=O)C1=CC=C2C=CC=CC2=C1

Specifications

Quantity 100 g
UN Number UN2585
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 208.23
Percent Purity 98%
Chemical Name or Material Naphthalene-2-sulfonic acid
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: ARYL SULFONIC ACIDS, SOLID
EINECSNumber 204-375-3
RTECSNumber QK1225000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only