missing translation for 'onlineSavingsMsg'
Learn More

Naphthalene-1-sulfonic acid sodium salt, 98% (dry wt.), water <3.5%

CAS: 130-14-3 | C10H7NaO3S | 230.213 g/mol

Supplier:  Thermo Scientific Chemicals B2248814

 View more versions of this product

Catalog No. AAB2248814


Only null left
Explore available promotions

Description

Description

Naphthalene-1-sulfonic acid sodium salt is used as a detection reagent to investigate ion-pair high-performance liquid chromatographic retention behavior of copper(II)-1-oxa-4,7,10,13-tetraazacyclopentadecane complex. It is also used to prepare naphthalene-1-sulfonyl chloride by reaction with phosphorousoxy chloride. Further, it is used as a stationary liquid phase for resolution of dichlorophenol isomers by gas chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Naphthalene-1-sulfonic acid sodium salt is used as a detection reagent to investigate ion-pair high-performance liquid chromatographic retention behavior of copper(II)-1-oxa-4,7,10,13-tetraazacyclopentadecane complex. It is also used to prepare naphthalene-1-sulfonyl chloride by reaction with phosphorousoxy chloride. Further, it is used as a stationary liquid phase for resolution of dichlorophenol isomers by gas chromatography.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.
Specifications

Specifications

Naphthalene-1-sulfonic acid sodium salt
299°C to 301°C
C10H7NaO3S
25 g
Hygroscopic
Soluble in water.
C1=CC=C2C(=C1)C=CC=C2S(=O)(=O)[O-].[Na+]
230.213
230.22
130-14-3
(dry wt.), water <2%
MFCD00064964
3639038
sodium 1-naphthalenesulfonate, sodium naphthalene-1-sulfonate, 1-naphthalenesulfonic acid sodium salt, unii-mai7v3c3pn, sodium naphthalenesulphonate, sodium naphthalene sulfonate, 1-naphthalenesulfonic acid, sodium salt, alpha salt, sodium alpha-naphthalenesulfonate, mai7v3c3pn
HIEHAIZHJZLEPQ-UHFFFAOYSA-M
sodium;naphthalene-1-sulfonate
23661867
99%

Chemical Identifiers

130-14-3
230.213
HIEHAIZHJZLEPQ-UHFFFAOYSA-M
23661867
C1=CC=C2C(=C1)C=CC=C2S(=O)(=O)[O-].[Na+]
C10H7NaO3S
MFCD00064964
sodium 1-naphthalenesulfonate, sodium naphthalene-1-sulfonate, 1-naphthalenesulfonic acid sodium salt, unii-mai7v3c3pn, sodium naphthalenesulphonate, sodium naphthalene sulfonate, 1-naphthalenesulfonic acid, sodium salt, alpha salt, sodium alpha-naphthalenesulfonate, mai7v3c3pn
sodium;naphthalene-1-sulfonate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P264b-P280i-P305+P351+P338

H319

EINECSNumber : 204-976-0

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only