missing translation for 'onlineSavingsMsg'
Learn More

Naphthalene-1,4-dicarboxylic acid, 98+%

CAS: 605-70-9 | C12H8O4 | 216.19 g/mol

Supplier:  Thermo Scientific Chemicals L2024922

 Afficher plus de versions de ce produit

Numéro de catalogue. AAL2024922


Il en reste null
Découvrez plus d'offres spéciales

Description

Description

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Insoluble in water.

Notes
Store away from water/moisture. Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Spécifications

Spécifications

Naphthalene-1,4-dicarboxylic acid
321°C to 325°C
MFCD00014312
2051255
Insoluble in water.
OC(=O)C1=C2C=CC=CC2=C(C=C1)C(O)=O
216.19
216.19
605-70-9
C12H8O4
100 g
1,4-naphthalenedicarboxylic acid, 1,4-naphthalenedicarboxylicacid, 1,4-naphthalicacid, 1,4-naphthalic acid, pubchem21317, acmc-1b0c7, ksc357a6b, naphthalene-1,4-dicarboxylicacid, naphthalene-1,4-dicarboxlic acid, naphthaline-1,4-dicarboxylic acid
ABMFBCRYHDZLRD-UHFFFAOYSA-N
naphthalene-1,4-dicarboxylic acid
69065
≥98%

Identifiants chimiques

605-70-9
216.19
ABMFBCRYHDZLRD-UHFFFAOYSA-N
69065
OC(=O)C1=C2C=CC=CC2=C(C=C1)C(O)=O
C12H8O4
MFCD00014312
1,4-naphthalenedicarboxylic acid, 1,4-naphthalenedicarboxylicacid, 1,4-naphthalicacid, 1,4-naphthalic acid, pubchem21317, acmc-1b0c7, ksc357a6b, naphthalene-1,4-dicarboxylicacid, naphthalene-1,4-dicarboxlic acid, naphthaline-1,4-dicarboxylic acid
naphthalene-1,4-dicarboxylic acid
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
Sécurité et manipulation

Sécurité et manipulation

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

missing translation for 'einecsNumber' : 210-094-7

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

FDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only