Learn More
Naphthalene-1,4,5,8-tetracarboxylic acid dianhydride, 97%
CAS: 81-30-1 | C14H4O6 | 268.18 g/mol
Supplier: Thermo Scientific Chemicals L1436514
Description
Naphthalene-1,4,5,8-tetracarboxylic acid dianhydride is used as a reagent in the preparation of novel fluorescent markers for hypoxic cells of naphthalimides with two heterocyclic side chains for bioreductive binding.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsNaphthalene-1,4,5,8-tetracarboxylic acid dianhydride is used as a reagent in the preparation of novel fluorescent markers for hypoxic cells of naphthalimides with two heterocyclic side chains for bioreductive binding.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. It is sensitive to moisture. Store under inert gas. Incompatible with acids, bases and oxidizing agents.
Specifications
| Naphthalene-1,4,5,8-tetracarboxylic acid dianhydride | |
| >300°C | |
| MFCD00006915 | |
| 272788 | |
| 1,4,5,8-naphthalenetetracarboxylic dianhydride, ntcda, naphthalenetetracarboxylic dianhydride, isochromeno 6,5,4-def isochromene-1,3,6,8-tetraone, ntda, unii-l56xqd1v6y, 2,7-dioxapyrene-1,3,6,8-tetrone, naphthalene-1,8:4,5-tetracarboxylic dianhydride, 1,8:4,5-naphthalenetetracarboxylic dianhydride, naphthalene-1,4,5,8-tetracarboxylic dianhydride | |
| C1=CC2=C3C(=CC=C4C3=C1C(=O)OC4=O)C(=O)OC2=O | |
| 6678 | |
| 97% |
| 81-30-1 | |
| C14H4O6 | |
| 25 g | |
| Moisture sensitive | |
| YTVNOVQHSGMMOV-UHFFFAOYSA-N | |
| 268.18 | |
| 268.18 |
Chemical Identifiers
| 81-30-1 | |
| 268.18 | |
| YTVNOVQHSGMMOV-UHFFFAOYSA-N | |
| 6678 |
| C14H4O6 | |
| MFCD00006915 | |
| 1,4,5,8-naphthalenetetracarboxylic dianhydride, ntcda, naphthalenetetracarboxylic dianhydride, isochromeno 6,5,4-def isochromene-1,3,6,8-tetraone, ntda, unii-l56xqd1v6y, 2,7-dioxapyrene-1,3,6,8-tetrone, naphthalene-1,8:4,5-tetracarboxylic dianhydride, 1,8:4,5-naphthalenetetracarboxylic dianhydride, naphthalene-1,4,5,8-tetracarboxylic dianhydride | |
| C1=CC2=C3C(=CC=C4C3=C1C(=O)OC4=O)C(=O)OC2=O |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 201-342-5
RTECSNumber : QK3695000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only