missing translation for 'onlineSavingsMsg'
Learn More

Naphthalene-1,4,5,8-tetracarboxylic acid dianhydride, 97%

Catalog No. p-7046451
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1436506 5 g
AAL1436514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1436506 Supplier Thermo Scientific Chemicals Supplier No. L1436506
Only null left

CAS: 81-30-1 | C14H4O6 | 268.18 g/mol

Naphthalene-1,4,5,8-tetracarboxylic acid dianhydride is used as a reagent in the preparation of novel fluorescent markers for hypoxic cells of naphthalimides with two heterocyclic side chains for bioreductive binding.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Naphthalene-1,4,5,8-tetracarboxylic acid dianhydride is used as a reagent in the preparation of novel fluorescent markers for hypoxic cells of naphthalimides with two heterocyclic side chains for bioreductive binding.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. It is sensitive to moisture. Store under inert gas. Incompatible with acids, bases and oxidizing agents.

Chemical Identifiers

CAS 81-30-1
Molecular Formula C14H4O6
Molecular Weight (g/mol) 268.18
MDL Number MFCD00006915
InChI Key YTVNOVQHSGMMOV-UHFFFAOYSA-N
Synonym 1,4,5,8-naphthalenetetracarboxylic dianhydride, ntcda, naphthalenetetracarboxylic dianhydride, isochromeno 6,5,4-def isochromene-1,3,6,8-tetraone, ntda, unii-l56xqd1v6y, 2,7-dioxapyrene-1,3,6,8-tetrone, naphthalene-1,8:4,5-tetracarboxylic dianhydride, 1,8:4,5-naphthalenetetracarboxylic dianhydride, naphthalene-1,4,5,8-tetracarboxylic dianhydride
PubChem CID 6678
SMILES C1=CC2=C3C(=CC=C4C3=C1C(=O)OC4=O)C(=O)OC2=O

Specifications

Melting Point >300°C
Quantity 5 g
Beilstein 272788
Sensitivity Moisture sensitive
Formula Weight 268.18
Percent Purity 97%
Chemical Name or Material Naphthalene-1,4,5,8-tetracarboxylic acid dianhydride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 201-342-5
RTECSNumber QK3695000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only