missing translation for 'onlineSavingsMsg'
Learn More

Naphthacene 97.0+%, TCI America™

Catalog No. p-7133565
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N0001100MG 100 mg
N00011G 1 g
N00015G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. N0001100MG Supplier TCI America Supplier No. N0001100MG
Only null left

Chemical Identifiers

CAS 92-24-0
Molecular Formula C18H12
Molecular Weight (g/mol) 228.294
MDL Number MFCD00003702
InChI Key IFLREYGFSNHWGE-UHFFFAOYSA-N
Synonym naphthacene, 2,3-benzanthracene, rubene, benz b anthracene, 2,3-benzanthrene, chrysogen, hydrocarbon, unii-qyj5z6712r, ccris 1183
PubChem CID 7080
ChEBI CHEBI:32600
IUPAC Name tetracene
SMILES C1=CC=C2C=C3C=C4C=CC=CC4=CC3=CC2=C1

Specifications

Melting Point 357°C
Quantity 100 mg
Formula Weight 228.29
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Naphthacene
RTECSNumber QI7605000
TSCA Yes