missing translation for 'onlineSavingsMsg'
Learn More

Naphthacene (purified by sublimation) 98.0+%, TCI America™

Catalog No. p-7134043
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N0951200MG 200 mg
N09511G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N0951200MG Supplier TCI America Supplier No. N0951200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 92-24-0
Molecular Formula C18H12
Molecular Weight (g/mol) 228.294
MDL Number MFCD00003702
InChI Key IFLREYGFSNHWGE-UHFFFAOYSA-N
Synonym naphthacene, 2,3-benzanthracene, rubene, benz b anthracene, 2,3-benzanthrene, chrysogen, hydrocarbon, unii-qyj5z6712r, ccris 1183
PubChem CID 7080
ChEBI CHEBI:32600
IUPAC Name tetracene
SMILES C1=CC=C2C=C3C=C4C=CC=CC4=CC3=CC2=C1

Specifications

Melting Point 357°C
Quantity 200 mg
Formula Weight 228.29
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Naphthacene (purified by sublimation)
RTECSNumber QI7605000
TSCA Yes