missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NANOS1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113725
Description
Immunogen sequence: DDDDDSDEPG SRGRYLGSAL ELRALELCAG PAEAGLLEER FAELSPFAGR For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Human: 100%; Mouse: 93%; Pig: 86%; Rabbit: 86%; Rat: 100%.
NANOS1 may regulate translation of specific mRNAs by forming a complex with PUM2 that associates with the 3'-UTR of mRNA targets.Chemical Identifiers
| NANOS1 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Polyclonal | |
| Liquid | |
| IgG | |
| Human | |
| NANOS1 | |
| EC_Rep1a; nanos 1; nanos C2HC-type zinc finger 1; nanos homolog 1; nanos homolog 1 (Drosophila); Nanos1; Nos; NOS1; NOS-1; RP11-498J9.3; SPGF12; ZC2HC12A | |
| NANOS1 | |
| Primary | |
| Affinity Chromatography |
| 0.5 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8WY41 | |
| 340719 | |
| Synthetic peptide directed towards the middle region of human NANOS1. | |
| Antibody |
Specifications
| NANOS1 | |
| Polyclonal | |
| Unconjugated | |
| NANOS1 | |
| EC_Rep1a; nanos 1; nanos C2HC-type zinc finger 1; nanos homolog 1; nanos homolog 1 (Drosophila); Nanos1; Nos; NOS1; NOS-1; RP11-498J9.3; SPGF12; ZC2HC12A | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 340719 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8WY41 | |
| NANOS1 | |
| Synthetic peptide directed towards the middle region of human NANOS1. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |