missing translation for 'onlineSavingsMsg'
Learn More

Nanodiamond (particle size : <10nm) 97.0+%, TCI America™

Catalog No. p-7134051
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N09621G 1 g
N09625G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N09621G Supplier TCI America Supplier No. N09621G
Only null left

Chemical Identifiers

CAS 7782-40-3
Molecular Formula C
Molecular Weight (g/mol) 12.011
MDL Number MFCD00211867
InChI Key OKTJSMMVPCPJKN-UHFFFAOYSA-N
PubChem CID 5462310
ChEBI CHEBI:27594
IUPAC Name carbon
SMILES [C]

Specifications

Color Gray-White
Quantity 1 g
Percent Purity ≥97.00%
Physical Form Powder
Chemical Name or Material Nanodiamond (particle size : <10nm)
RTECSNumber HL4158550
TSCA Yes