CAS RN 7782-40-3
CAS RN 7782-40-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Nanodiamond (particle size : <10nm) (5%, Dispersion in Water), TCI America™
CAS: 7782-40-3 Molecular Formula: C Molecular Weight (g/mol): 12.011 MDL Number: MFCD00211867 InChI Key: OKTJSMMVPCPJKN-UHFFFAOYSA-N PubChem CID: 5462310 ChEBI: CHEBI:27594 IUPAC Name: carbon SMILES: [C]
2
Nanodiamond (particle size : <10nm) 97.0+%, TCI America™
CAS: 7782-40-3 Molecular Formula: C Molecular Weight (g/mol): 12.011 MDL Number: MFCD00211867 InChI Key: OKTJSMMVPCPJKN-UHFFFAOYSA-N PubChem CID: 5462310 ChEBI: CHEBI:27594 IUPAC Name: carbon SMILES: [C]