Learn More
Promega NanoBRET TE Intracellular Kinase Assay, K-5, 100 assays
The NanoBRET Target Engagement (TE) Assay measures compound binding at select kinase targets within intact cells. The assay is based on the NanoBRET System, an energy transfer technique designed to measure molecular proximity in living cells.
Supplier: Promega N2500
Description
- Measure Compound Binding at Kinase Targets in Intact Cells
- Detect kinase target engagement in live cells
- Choose from >125 kinase fusion vectors
- See Kinase Table (below) for available kinases and related information
Specifications
| 100 Assays | |
| <−65°C |