missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Promega Nano-Glo™ Luciferase Assay System
The Nano-Glo™ Luciferase Assay System provides a simple, single-addition reagent that generates a glow-type signal in the presence of NanoLuc luciferase with a half-life of approximately 120 minutes in commonly used tissue culture media.
$332.04 - $14996.92
Specifications
| Type | Nano-Glo Luciferase Assay Buffer |
|---|---|
| Content And Storage | −30 to −10°C |
| For Use With (Application) | Provides a single addition reagent that generates a glow-type signal in the presence of NanoLuc™ luciferase |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
PRN1150
|
Promega
N1150 |
10 x 100 mL |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
PRN1110
|
Promega
N1110 |
10 mL |
Each for $332.04
|
|
|||||
PRN1130
|
Promega
N1130 |
10 x 10 mL |
Each for $2,341.37
|
|
|||||
PRN1120
|
Promega
N1120 |
100 mL |
Each for $1,936.87
|
|
|||||
Description
- Add-Mix-Measure assay for NanoLuc(R) Luciferase
- Bright NanoLuc(R) reporter enables use in applications where sensitivity is limited
- Add-and-read simplicity allows scaling from bench to HTS
- Extended bright light output optimized for batch and continuous processing
Specifications
| Nano-Glo Luciferase Assay Buffer | |
| −30 to −10°C | |
| Provides a single addition reagent that generates a glow-type signal in the presence of NanoLuc™ luciferase |