missing translation for 'onlineSavingsMsg'
Learn More

Promega Nano-Glo™ Luciferase Assay System

Catalog No. PRN1150
Encompass
Change view
Click to view available options
Quantity:
10 mL
100 mL
10 x 10 mL
10 x 100 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRN1150 10 x 100 mL
PRN1110 10 mL
PRN1130 10 x 10 mL
PRN1120 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. PRN1150 Supplier Promega Supplier No. N1150

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The Nano-Glo™ Luciferase Assay System provides a simple, single-addition reagent that generates a glow-type signal in the presence of NanoLuc luciferase with a half-life of approximately 120 minutes in commonly used tissue culture media.

  • Add-Mix-Measure assay for NanoLuc(R) Luciferase
  • Bright NanoLuc(R) reporter enables use in applications where sensitivity is limited
  • Add-and-read simplicity allows scaling from bench to HTS
  • Extended bright light output optimized for batch and continuous processing

Specifications

Kit Contents 5 x 4mL Nano-Glo™ Luciferase assay Substrate, 10 x 100mL Nano-Glo™ Luciferase Assay Buffer, integral Lysis Buffer reagent
Type Nano-Glo Luciferase Assay Buffer
Quantity 10 x 100 mL
Content And Storage −30 to −10°C
For Use With (Application) Provides a single addition reagent that generates a glow-type signal in the presence of NanoLuc™ luciferase