missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Promega Nano-Glo Live Cell Assay System
The Nano-Glo™ Live Cell Assay System is a single-addition, non-lytic detection reagent used to measure NanoBiT or NanoLuc luminescence from living cells.
$341.25 - $18,232.95
Specifications
| Content And Storage | -30°C to -10°C |
|---|---|
| For Use With (Application) | NanoBiT™ protein complementation or NanoLuc™ reporter activity |
| Includes | Substrate and Buffer |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PRN2011
|
Promega
N2011 |
100 Assays |
Each for $341.25
|
|
|||||
PRN2012
|
Promega
N2012 |
1000 Assays |
Each for $2,042.28
|
|
|||||
PRN2013
|
Promega
N2013 |
10,000 Assays |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
- Measures NanoLuc(R) Luminescence in Living Cells
- Substrate and buffer optimized to enhance stability
- Use to detect NanoBiT(R) protein complementation or NanoLuc(R) reporter activity
- Monitors luminescence at a single time point or continuously for up to 2 hours without compromising cell viability.
Specifications
| -30°C to -10°C | |
| NanoBiT™ protein complementation or NanoLuc™ reporter activity | |
| Substrate and Buffer |