missing translation for 'onlineSavingsMsg'
Learn More

NAMPT/PBEF/Visfatin Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501558423
Encompass
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
501558423 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 501558423 Supplier Bethyl Laboratories Inc Supplier No. A300779A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

Visfatin, also known as pre-B cell colony-enhancing factor, is an adipocytokine that is abundantly secreted by visceral adipose tissue, with insulin mimetic effects. Visfatin mimics insulin signaling by binding to the insulin receptor and reducing glucose release from liver cells, while stimulating glucose utilization in adipocytes and myocytes. Visfatin is also invoved in enhancement of cell proliferation, biosynthesis of nicotinamide mono-and dinucleotide and glucose homeostasis.

Specifications

Antigen NAMPT/PBEF/Visfatin
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.2 mg/ml
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene NAMPT
Gene Accession No. P43490
Gene Alias NAMPT, Nicotinamide phosphoribosyltransferase, EC 2.4.2.12, NAmPRTase, Nampt, Pre-B cell-enhancing factor, Pre-B-cell colony-enhancing factor 1
Gene Symbols NAMPT
Host Species Rabbit
Immunogen Between 450 and C-term
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10135
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less