missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NAM 98.0+%, TCI America™
$326.21 - $585.32
Chemical Identifiers
| CAS | 49759-20-8 |
|---|---|
| Molecular Formula | C17H10N2O2 |
| Molecular Weight (g/mol) | 274.279 |
| MDL Number | MFCD00042983 |
| InChI Key | NCFIKBMPEOEIED-UHFFFAOYSA-N |
| Synonym | N-(9-Acridinyl)maleimide, 9-Maleimidoacridine |
| PubChem CID | 3016496 |
| IUPAC Name | 1-acridin-9-ylpyrrole-2,5-dione |
| SMILES | C1=CC=C2C(=C1)C(=C3C=CC=CC3=N2)N4C(=O)C=CC4=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
A559150MG
|
TCI America
A559150MG |
50 mg |
Each for $326.21
|
|
|||||
|
A5591100MG
|
TCI America
A5591100MG |
100 mg |
Each for $585.32
|
|
|||||
Chemical Identifiers
| 49759-20-8 | |
| 274.279 | |
| NCFIKBMPEOEIED-UHFFFAOYSA-N | |
| 3016496 | |
| C1=CC=C2C(=C1)C(=C3C=CC=CC3=N2)N4C(=O)C=CC4=O |
| C17H10N2O2 | |
| MFCD00042983 | |
| N-(9-Acridinyl)maleimide, 9-Maleimidoacridine | |
| 1-acridin-9-ylpyrrole-2,5-dione |
Specifications
| 49759-20-8 | |
| C17H10N2O2 | |
| 50 mg | |
| NCFIKBMPEOEIED-UHFFFAOYSA-N | |
| 1-acridin-9-ylpyrrole-2,5-dione | |
| 3016496 | |
| ≥98.0% (HPLC) | |
| NAM [=N-(9-Acridinyl)maleimide] [for HPLC Labeling] |
| Yellow | |
| MFCD00042983 | |
| N-(9-Acridinyl)maleimide, 9-Maleimidoacridine | |
| C1=CC=C2C(=C1)C(=C3C=CC=CC3=N2)N4C(=O)C=CC4=O | |
| 274.279 | |
| 274.28 | |
| Crystalline Powder |
Safety and Handling
TSCA : No