accessibility menu, dialog, popup

UserName

CAS RN 49759-20-8

IUPAC Name: 1-(acridin-9-yl)-2,5-dihydro-1H-pyrrole-2,5-dione
Synonyms: 1-(acridin-9-yl)pyrrole-2,5-dione
Molecular Weight (g/mol): 274.28
Molecular Formula: C17H10N2O2
InChi Key: NCFIKBMPEOEIED-UHFFFAOYSA-N
SMILES: O=C1C=CC(=O)N1C1=C2C=CC=CC2=NC2=CC=CC=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results