missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NAM 98.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Quantity | 50 mg |
| Formula Weight | 274.28 |
| Percent Purity | ≥98.0% (HPLC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | NAM [=N-(9-Acridinyl)maleimide] [for HPLC Labeling] |
Safety and Handling
| TSCA | No |