missing translation for 'onlineSavingsMsg'
Learn More

Naltrexone hydrochloride, Thermo Scientific Chemicals

A congener of naloxone and also an opioid antagonist

Supplier:  Thermo Scientific Chemicals J60590MD

 View more versions of this product

Catalog No. AAJ60590MD


Only null left
Explore available promotions

Description

Description

A congener of naloxone and also an opioid antagonist.Naltrexone hydrochloride is used in the treatment of alcohol and utilized for deceasing drinking habit. It is also used to prevent euphorigenic effects in the treatment of patients addicted to opioids. Further, it is used as a specific opiate antagonist.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A congener of naloxone and also an opioid antagonist.Naltrexone hydrochloride is used in the treatment of alcohol and utilized for deceasing drinking habit. It is also used to prevent euphorigenic effects in the treatment of patients addicted to opioids. Further, it is used as a specific opiate antagonist.

Solubility
Soluble in water at 50mg/ml.

Notes
Light sensitive. Incompatible with bases and oxidizing agents. Store in a cool place.
Greener Choice Claims
  • Environmental benefits include:
    • Renewable Energy
Specifications
Show More
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302
Harmful if swallowed.

P264b-P270-P301+P312-P330-P501c

H302

EINECSNumber : 240-723-0

RTECSNumber : QD2160000

TSCA : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only