missing translation for 'onlineSavingsMsg'
Learn More

Naltrexone hydrochloride, Thermo Scientific Chemicals

Catalog No. p-7041883
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ60590MD 250 mg
AAJ6059003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ60590MD Supplier Thermo Scientific Chemicals Supplier No. J60590MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A congener of naloxone and also an opioid antagonist

A congener of naloxone and also an opioid antagonist.Naltrexone hydrochloride is used in the treatment of alcohol and utilized for deceasing drinking habit. It is also used to prevent euphorigenic effects in the treatment of patients addicted to opioids. Further, it is used as a specific opiate antagonist.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A congener of naloxone and also an opioid antagonist.Naltrexone hydrochloride is used in the treatment of alcohol and utilized for deceasing drinking habit. It is also used to prevent euphorigenic effects in the treatment of patients addicted to opioids. Further, it is used as a specific opiate antagonist.

Solubility
Soluble in water at 50mg/ml.

Notes
Light sensitive. Incompatible with bases and oxidizing agents. Store in a cool place.

Chemical Identifiers

CAS 16676-29-2
Molecular Formula C20H24ClNO4
Molecular Weight (g/mol) 377.865
MDL Number MFCD00069324
InChI Key RHBRMCOKKKZVRY-ITLPAZOVSA-N
Synonym naltrexone hydrochloride, naltrexone hcl, trexan, depade, antaxone, revia, nalorex, unii-z6375yw9sf, nemexin
PubChem CID 5485201
IUPAC Name (4R,4aS,7aR,12bS)-3-(cyclopropylmethyl)-4a,9-dihydroxy-2,4,5,6,7a,13-hexahydro-1H-4,12-methanobenzofuro[3,2-e]isoquinoline-7-one;hydrochloride
SMILES C1CC1CN2CCC34C5C(=O)CCC3(C2CC6=C4C(=C(C=C6)O)O5)O.Cl

Specifications

Color White
Quantity 250 mg
Beilstein 3580333
Sensitivity Light sensitive
Merck Index 14,6363
Solubility Information Soluble in water at 50mg/ml.
Formula Weight 377.86
Physical Form Powder
Chemical Name or Material Naltrexone hydrochloride
Hazard Category H302
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 240-723-0
RTECSNumber QD2160000
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only