missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nalpha-(tert-Butoxycarbonyl)-Nepsilon-carbobenzoxy-L-lysine 98.0+%, TCI America™
$37.97 - $113.92
Chemical Identifiers
| CAS | 2389-45-9 |
|---|---|
| Molecular Formula | C19H28N2O6 |
| Molecular Weight (g/mol) | 380.44 |
| MDL Number | MFCD00065584 |
| InChI Key | BDHUTRNYBGWPBL-GGYSOQFKNA-N |
| Synonym | boc-lys z-oh, n-boc-n'-cbz-l-lysine, boc-lys cbz-oh, boc-l-lys z-oh, nalpha-boc-nepsilon-cbz-l-lysine, s-6-benzyloxy carbonyl amino-2-tert-butoxycarbonyl amino hexanoic acid, 2s-6-benzyloxy carbonyl amino-2-tert-butoxycarbonyl amino hexanoic acid, n-alpha-boc-n-epsilon-benzyloxycarbonyl-l-lysine, nalpha-tert-butoxycarbonyl-nepsilon-carbobenzoxy-l-lysine, n-alpha-t-butyloxycarbonyl-n-epsilon-benzyloxycarbonyl-l-lysine |
| PubChem CID | 2724765 |
| IUPAC Name | (2S)-6-{[(benzyloxy)carbonyl]amino}-2-{[(tert-butoxy)carbonyl]amino}hexanoic acid |
| SMILES | CC(C)(C)OC(=O)N[C@@H](CCCCNC(=O)OCC1=CC=CC=C1)C(O)=O |
Chemical Identifiers
| 2389-45-9 | |
| 380.44 | |
| BDHUTRNYBGWPBL-GGYSOQFKNA-N | |
| 2724765 | |
| CC(C)(C)OC(=O)N[C@@H](CCCCNC(=O)OCC1=CC=CC=C1)C(O)=O |
| C19H28N2O6 | |
| MFCD00065584 | |
| boc-lys z-oh, n-boc-n'-cbz-l-lysine, boc-lys cbz-oh, boc-l-lys z-oh, nalpha-boc-nepsilon-cbz-l-lysine, s-6-benzyloxy carbonyl amino-2-tert-butoxycarbonyl amino hexanoic acid, 2s-6-benzyloxy carbonyl amino-2-tert-butoxycarbonyl amino hexanoic acid, n-alpha-boc-n-epsilon-benzyloxycarbonyl-l-lysine, nalpha-tert-butoxycarbonyl-nepsilon-carbobenzoxy-l-lysine, n-alpha-t-butyloxycarbonyl-n-epsilon-benzyloxycarbonyl-l-lysine | |
| (2S)-6-{[(benzyloxy)carbonyl]amino}-2-{[(tert-butoxy)carbonyl]amino}hexanoic acid |
Specifications
| 2389-45-9 | |
| White | |
| MFCD00065584 | |
| boc-lys z-oh, n-boc-n'-cbz-l-lysine, boc-lys cbz-oh, boc-l-lys z-oh, nalpha-boc-nepsilon-cbz-l-lysine, s-6-benzyloxy carbonyl amino-2-tert-butoxycarbonyl amino hexanoic acid, 2s-6-benzyloxy carbonyl amino-2-tert-butoxycarbonyl amino hexanoic acid, n-alpha-boc-n-epsilon-benzyloxycarbonyl-l-lysine, nalpha-tert-butoxycarbonyl-nepsilon-carbobenzoxy-l-lysine, n-alpha-t-butyloxycarbonyl-n-epsilon-benzyloxycarbonyl-l-lysine | |
| CC(C)(C)OC(=O)N[C@@H](CCCCNC(=O)OCC1=CC=CC=C1)C(O)=O | |
| 380.44 | |
| 380.44 | |
| Crystalline Powder |
| 77°C | |
| C19H28N2O6 | |
| 5 g | |
| BDHUTRNYBGWPBL-GGYSOQFKNA-N | |
| (2S)-6-{[(benzyloxy)carbonyl]amino}-2-{[(tert-butoxy)carbonyl]amino}hexanoic acid | |
| 2724765 | |
| ≥98.0% (HPLC,T) | |
| Nalpha-(tert-Butoxycarbonyl)-Nepsilon-carbobenzoxy-L-lysine |
Safety and Handling
TSCA : Yes