missing translation for 'onlineSavingsMsg'
Learn More

N-alpha-(tert-Butoxycarbonyl)-L-tryptophanol 98.0+%, TCI America™

Catalog No. p-7123446
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B42871G 1 g
B42875G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B42871G Supplier TCI America Supplier No. B42871G
Only null left

Chemical Identifiers

CAS 82689-19-8
Molecular Formula C16H22N2O3
Molecular Weight (g/mol) 290.36
MDL Number MFCD00235953
InChI Key JEFQUFUAEKORKL-LBPRGKRZSA-N
Synonym n-boc-l-tryptophanol, boc-tryptophanol, boc-l-tryptophanol, boc-trp-ol, s-tert-butyl 1-hydroxy-3-1h-indol-3-yl propan-2-yl carbamate, s-3-2-boc-amino-3-hydroxypropyl-indole, boc-d-trp-ol, n-alpha-boc-l-tryptophanol, boc-trypotophanol, boc-l-trp-ol
PubChem CID 7019533
IUPAC Name tert-butyl N-[(2S)-1-hydroxy-3-(1H-indol-3-yl)propan-2-yl]carbamate
SMILES CC(C)(C)OC(=O)N[C@H](CO)CC1=CNC2=CC=CC=C12

Specifications

Melting Point 122°C
Color White
Quantity 1 g
Formula Weight 290.36
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material N-alpha-(tert-Butoxycarbonyl)-L-tryptophanol