accessibility menu, dialog, popup

UserName

NH(S)HNOHOOCH3CH3CH3

CAS RN 82689-19-8

IUPAC Name: tert-butyl N-[(2S)-1-hydroxy-3-(1H-indol-3-yl)propan-2-yl]carbamate
Synonyms: tert-butyl N-[(2S)-1-hydroxy-3-(1H-indol-3-yl)propan-2-yl]carbamate|N-BOC-L-TRYPTOPHANOL
Molecular Weight (g/mol): 290.36
Molecular Formula: C16H22N2O3
InChi Key: JEFQUFUAEKORKL-LBPRGKRZSA-N
SMILES: CC(C)(C)OC(=O)N[C@H](CO)CC1=CNC2=CC=CC=C12
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)