missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nalpha-(tert-Butoxycarbonyl)-D-lysine 98.0+%, TCI America™
$54.06 - $212.51
Chemical Identifiers
| CAS | 106719-44-2 |
|---|---|
| Molecular Formula | C11H22N2O4 |
| Molecular Weight (g/mol) | 246.31 |
| MDL Number | MFCD00076956 |
| InChI Key | DQUHYEDEGRNAFO-SVGMAFHSNA-N |
| Synonym | boc-d-lys-oh, n-alpha-t-butoxycarbonyl-d-lysine, boc-l-lys-oh, nalpha-tert-butoxycarbonyl-d-lysine, nalpha-tert-butoxycarbonyl-d-lysine, n-, a-boc-d-lysine, boc-d-lysine, boc-d-lysine, n-tert-butoxycarbonyl-d-lysine, n-tert-butoxycarbonyl-d-lysine |
| PubChem CID | 7018770 |
| IUPAC Name | (2R)-6-amino-2-{[(tert-butoxy)carbonyl]amino}hexanoic acid |
| SMILES | CC(C)(C)OC(=O)N[C@H](CCCCN)C(O)=O |
Chemical Identifiers
| 106719-44-2 | |
| 246.31 | |
| DQUHYEDEGRNAFO-SVGMAFHSNA-N | |
| 7018770 | |
| CC(C)(C)OC(=O)N[C@H](CCCCN)C(O)=O |
| C11H22N2O4 | |
| MFCD00076956 | |
| boc-d-lys-oh, n-alpha-t-butoxycarbonyl-d-lysine, boc-l-lys-oh, nalpha-tert-butoxycarbonyl-d-lysine, nalpha-tert-butoxycarbonyl-d-lysine, n-, a-boc-d-lysine, boc-d-lysine, boc-d-lysine, n-tert-butoxycarbonyl-d-lysine, n-tert-butoxycarbonyl-d-lysine | |
| (2R)-6-amino-2-{[(tert-butoxy)carbonyl]amino}hexanoic acid |
Specifications
| 106719-44-2 | |
| White | |
| MFCD00076956 | |
| boc-d-lys-oh, n-alpha-t-butoxycarbonyl-d-lysine, boc-l-lys-oh, nalpha-tert-butoxycarbonyl-d-lysine, nalpha-boc-d-lysine, nalpha-boc-d-lysine, a-boc-d-lysine, boc-d-lysine, d-lysine, n2-1,1-dimethylethoxy carbonyl, d-lysine, n2-1,1-dimethylethoxy carbonyl, n-tert-butoxycarbonyl-d-lysine | |
| CC(C)(C)OC(=O)N[C@H](CCCCN)C(O)=O | |
| 246.31 | |
| 246.31 | |
| Crystalline Powder |
| 198°C | |
| C11H22N2O4 | |
| 1 g | |
| DQUHYEDEGRNAFO-SVGMAFHSNA-N | |
| (2R)-6-amino-2-{[(tert-butoxy)carbonyl]amino}hexanoic acid | |
| 7018770 | |
| ≥98.0% (T) | |
| Nalpha-(tert-Butoxycarbonyl)-D-lysine |
Safety and Handling
TSCA : No