missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nalpha,Nepsilon-Di-Boc-L-lysine, 98%
CAS: 2483-46-7 | C16H30N2O6 | 346.42 g/mol
$134.75 - $407.38
Chemical Identifiers
| CAS | 2483-46-7 |
|---|---|
| Molecular Formula | C16H30N2O6 |
| Molecular Weight (g/mol) | 346.42 |
| MDL Number | MFCD00038515 |
| InChI Key | FBVSXKMMQOZUNU-LDGXTIHJNA-N |
| Synonym | boc-lys boc-oh, s-2,6-bis-tert-butoxycarbonylaminohexanoic acid, boc-lys boc, s-2,6-bis tert-butoxycarbonyl amino hexanoic acid, di-boc-l-lysine, na, ne-bis-boc-l-lysine, boc-l-lys boc-oh, boc-l-lys boc, boc-l-lysine boc, s-2,6-bis-tert-butoxycarbonylamino-hexanoic acid |
| PubChem CID | 7349648 |
| IUPAC Name | (2S)-2,6-bis[(2-methylpropan-2-yl)oxycarbonylamino]hexanoic acid |
| SMILES | CC(C)(C)OC(=O)NCCCC[C@H](NC(=O)OC(C)(C)C)C(O)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AAH6201914
|
N/A | 25 g |
Each for $134.75
|
|
|||||
|
AAH6201922
|
N/A | 100 g |
Each for $407.38
|
|
|||||
Description
Nalpha, Nepsilon-Di-Boc-L-lysine is used as fine chemical intermediate, synthesis intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsNalpha, Nepsilon-Di-Boc-L-lysine is used as fine chemical intermediate, synthesis intermediates.
Solubility
Soluble in chloroform, ether, ethyl acetate and methanol
Notes
Keep container tightly closed. Store away from oxidizing agents.
Chemical Identifiers
| 2483-46-7 | |
| 346.42 | |
| FBVSXKMMQOZUNU-LDGXTIHJNA-N | |
| 7349648 | |
| CC(C)(C)OC(=O)NCCCC[C@H](NC(=O)OC(C)(C)C)C(O)=O |
| C16H30N2O6 | |
| MFCD00038515 | |
| boc-lys boc-oh, s-2,6-bis-tert-butoxycarbonylaminohexanoic acid, boc-lys boc, s-2,6-bis tert-butoxycarbonyl amino hexanoic acid, di-boc-l-lysine, na, ne-bis-boc-l-lysine, boc-l-lys boc-oh, boc-l-lys boc, boc-l-lysine boc, s-2,6-bis-tert-butoxycarbonylamino-hexanoic acid | |
| (2S)-2,6-bis[(2-methylpropan-2-yl)oxycarbonylamino]hexanoic acid |
Specifications
| 2483-46-7 | |
| C16H30N2O6 | |
| boc-lys boc-oh, s-2,6-bis-tert-butoxycarbonylaminohexanoic acid, boc-lys boc, s-2,6-bis tert-butoxycarbonyl amino hexanoic acid, di-boc-l-lysine, na, ne-bis-boc-l-lysine, boc-l-lys boc-oh, boc-l-lys boc, boc-l-lysine boc, s-2,6-bis-tert-butoxycarbonylamino-hexanoic acid | |
| FBVSXKMMQOZUNU-LDGXTIHJNA-N | |
| (2S)-2,6-bis[(2-methylpropan-2-yl)oxycarbonylamino]hexanoic acid | |
| 7349648 | |
| 98% | |
| Nalpha,Nepsilon-Di-Boc-L-lysine |
| White | |
| MFCD00038515 | |
| Soluble in chloroform,ether,ethyl acetate and methanol | |
| CC(C)(C)OC(=O)NCCCC[C@H](NC(=O)OC(C)(C)C)C(O)=O | |
| 346.42 | |
| 346.42 | |
| Low Melting Solid |
Safety and Handling
TSCA : No
storageNote1 : -30° to -10°C
Recommended Storage : Store at -20°C
RUO – Research Use Only