missing translation for 'onlineSavingsMsg'
Learn More

Nalpha,Nepsilon-Di-Boc-L-lysine, 98%

Catalog No. AAH6201922
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6201922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6201922 Supplier Thermo Scientific Chemicals Supplier No. H6201922
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2483-46-7 | C16H30N2O6 | 346.42 g/mol

Nalpha, Nepsilon-Di-Boc-L-lysine is used as fine chemical intermediate, synthesis intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Nalpha, Nepsilon-Di-Boc-L-lysine is used as fine chemical intermediate, synthesis intermediates.

Solubility
Soluble in chloroform, ether, ethyl acetate and methanol

Notes
Keep container tightly closed. Store away from oxidizing agents.

Chemical Identifiers

CAS 2483-46-7
Molecular Formula C16H30N2O6
Molecular Weight (g/mol) 346.42
MDL Number MFCD00038515
InChI Key FBVSXKMMQOZUNU-LDGXTIHJNA-N
Synonym boc-lys boc-oh, s-2,6-bis-tert-butoxycarbonylaminohexanoic acid, boc-lys boc, s-2,6-bis tert-butoxycarbonyl amino hexanoic acid, di-boc-l-lysine, na, ne-bis-boc-l-lysine, boc-l-lys boc-oh, boc-l-lys boc, boc-l-lysine boc, s-2,6-bis-tert-butoxycarbonylamino-hexanoic acid
PubChem CID 7349648
IUPAC Name (2S)-2,6-bis[(2-methylpropan-2-yl)oxycarbonylamino]hexanoic acid
SMILES CC(C)(C)OC(=O)NCCCC[C@H](NC(=O)OC(C)(C)C)C(O)=O

Specifications

Color White
Solubility Information Soluble in chloroform,ether,ethyl acetate and methanol
Quantity 100 g
Formula Weight 346.42
Percent Purity 98%
Physical Form Low Melting Solid
Chemical Name or Material Nalpha,Nepsilon-Di-Boc-L-lysine
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only